Learn More
Medchemexpress LLC Dibutyl phenyl phosphate | 2528-36-1 | MFCD00438060 | 286.30 g/mol | C14H23O4P | 10 MG
SDP
Catalog No. 5000283232
Encompass_Preferred
Dibutyl phenyl phosphate is an organophosphate compound used as a flame retardant and a research reagent. It has reported butyrylcholinesterase (BChE) inhibitory activity (IC50: 83.6 μM; Ki: 94.5 μM) and is supplied in small milligram quantities for laboratory research; refer to the certificate of analysis for purity and storage details.
  • Organophosphate flame retardant.
  • Reported butyrylcholinesterase (BChE) inhibitor (IC50: 83.6 μM; Ki: 94.5 μM).
  • Molecular formula C14H23O4P and molecular weight 286.30 g/mol.
  • Supplied in small milligram quantities for research applications.
  • Certificate of analysis contains purity and storage instructions.
  • Intended for research use only; not for human use.
Catalog No. 50-002-83232 Supplier Medchemexpress LLC Supplier No. HYW18533610MG
Dibutyl phenyl phosphate is an organophosphate compound used as a flame retardant and a research reagent. It has reported butyrylcholinesterase (BChE) inhibitory activity (IC50: 83.6 μM; Ki: 94.5 μM) and is supplied in small milligram quantities for laboratory research; refer to the certificate of analysis for purity and storage details.
  • Organophosphate flame retardant.
  • Reported butyrylcholinesterase (BChE) inhibitor (IC50: 83.6 μM; Ki: 94.5 μM).
  • Molecular formula C14H23O4P and molecular weight 286.30 g/mol.
  • Supplied in small milligram quantities for research applications.
  • Certificate of analysis contains purity and storage instructions.
  • Intended for research use only; not for human use.