Learn More
Medchemexpress LLC Dibutyl phenyl phosphate | 2528-36-1 | MFCD00438060 | 286.30 g·mol⁻¹ | C14H23O4P | 50 MG
SDP
Catalog No. 5000283067
Encompass_Preferred
Dibutyl phenyl phosphate is an organophosphate research reagent used primarily as a flame retardant and reported to inhibit butyrylcholinesterase (BChE). It is supplied for laboratory research use only; consult product documentation for handling, purity, and storage requirements.
  • Used as a flame retardant in material testing and formulation studies.
  • Inhibits butyrylcholinesterase (BChE); IC50 83.6 μM, Ki 94.5 μM.
  • Documented molecular formula C14H23O4P and molecular weight 286.30 g·mol⁻¹.
  • Supplied in small pack sizes appropriate for research applications.
  • Certificate of analysis and supporting documentation available on request for purity and storage details.
Catalog No. 50-002-83067 Supplier Medchemexpress LLC Supplier No. HYW18533650MG
Dibutyl phenyl phosphate is an organophosphate research reagent used primarily as a flame retardant and reported to inhibit butyrylcholinesterase (BChE). It is supplied for laboratory research use only; consult product documentation for handling, purity, and storage requirements.
  • Used as a flame retardant in material testing and formulation studies.
  • Inhibits butyrylcholinesterase (BChE); IC50 83.6 μM, Ki 94.5 μM.
  • Documented molecular formula C14H23O4P and molecular weight 286.30 g·mol⁻¹.
  • Supplied in small pack sizes appropriate for research applications.
  • Certificate of analysis and supporting documentation available on request for purity and storage details.