Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibutyl phenyl phosphate | 2528-36-1 | MFCD00438060 | 99.0% | 286.30 g·mol⁻¹ | C14H23O4P | 5 MG
SDP

Catalog No. 5000281922
Encompass_Preferred

Dibutyl phenyl phosphate is an organophosphate research reagent used primarily as a flame retardant and studied for biological activity. It is reported to inhibit butyrylcholinesterase (BChE) with IC50 83.6 μM and Ki 94.5 μM. Intended for research use only and not for human or clinical applications.

  • Organophosphate flame retardant.
  • Reported BChE inhibitor (IC50 83.6 μM; Ki 94.5 μM).
  • Chemical formula C14H23O4P; molecular weight 286.30 g·mol⁻¹.
  • CAS number 2528-36-1 for unambiguous identification.
  • Supplied in milligram-scale quantities suitable for laboratory assays.
  • Intended for research use only; follow safety data sheet guidance for handling.

Catalog No. 50-002-81922 Supplier Medchemexpress LLC Supplier No. HYW1853365MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibutyl phenyl phosphate is an organophosphate research reagent used primarily as a flame retardant and studied for biological activity. It is reported to inhibit butyrylcholinesterase (BChE) with IC50 83.6 μM and Ki 94.5 μM. Intended for research use only and not for human or clinical applications.

  • Organophosphate flame retardant.
  • Reported BChE inhibitor (IC50 83.6 μM; Ki 94.5 μM).
  • Chemical formula C14H23O4P; molecular weight 286.30 g·mol⁻¹.
  • CAS number 2528-36-1 for unambiguous identification.
  • Supplied in milligram-scale quantities suitable for laboratory assays.
  • Intended for research use only; follow safety data sheet guidance for handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.