Learn More
Medchemexpress LLC Dibutyl phosphate-d18 | 156213-21-7 | MFCD00012206 | >95.0% | C8HD18O4P | 10MG
SDP
Catalog No. 5000261078
Encompass_Preferred
Dibutyl phosphate-d18 is a deuterium-labeled analogue of dibutyl phosphate (CAS 156213-21-7) used as a tracer and internal standard for quantitative analytical methods. The isotopic labeling enables accurate quantitation and method development in NMR, GC-MS, and LC-MS, and supports metabolite and trace-level studies.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS workflows.
  • High isotopic enrichment reduces interference from unlabeled analytes.
  • Provided as a stable liquid for easy handling and spiking.
  • Available in small milligram quantities for reference and method validation.
Catalog No. 50-002-61078 Supplier Medchemexpress LLC Supplier No. HY135604S10MG
Dibutyl phosphate-d18 is a deuterium-labeled analogue of dibutyl phosphate (CAS 156213-21-7) used as a tracer and internal standard for quantitative analytical methods. The isotopic labeling enables accurate quantitation and method development in NMR, GC-MS, and LC-MS, and supports metabolite and trace-level studies.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS workflows.
  • High isotopic enrichment reduces interference from unlabeled analytes.
  • Provided as a stable liquid for easy handling and spiking.
  • Available in small milligram quantities for reference and method validation.