Learn More
Medchemexpress LLC Dibutyl phosphate-d18 | 156213-21-7 | MFCD00012206 | 98.5% | 228.32 | C8HD18O4P | 5 MG
SDP
Catalog No. 5000223048
Encompass_Preferred
Dibutyl phosphate-d18 is a deuterium-labeled analogue of dibutyl phosphate provided as a small-mass analytical standard for research and analytical applications. It is intended for use as an isotopically labeled tracer or internal standard in mass spectrometry and method development, supplied at high purity for reliable analytical results.
  • Deuterium-labeled internal standard for analytical quantification.
  • High purity suitable for mass spectrometry and chromatographic methods.
  • Available in milligram-scale quantities for laboratory use.
  • Useful for tracer studies, method validation, and calibration.
  • CAS 156213-21-7 for unambiguous identification.
Catalog No. 50-002-23048 Supplier Medchemexpress LLC Supplier No. HY135604S5MG
Dibutyl phosphate-d18 is a deuterium-labeled analogue of dibutyl phosphate provided as a small-mass analytical standard for research and analytical applications. It is intended for use as an isotopically labeled tracer or internal standard in mass spectrometry and method development, supplied at high purity for reliable analytical results.
  • Deuterium-labeled internal standard for analytical quantification.
  • High purity suitable for mass spectrometry and chromatographic methods.
  • Available in milligram-scale quantities for laboratory use.
  • Useful for tracer studies, method validation, and calibration.
  • CAS 156213-21-7 for unambiguous identification.