Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl phosphite, 14.5-16% P

Catalog No. p-103413
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155061000 100 g Glass bottle
AC155065000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155061000 Supplier Thermo Scientific Chemicals Supplier No. 155061000
Only null left
Add to Cart

CAS: 1809-19-4 | C8H19O3P | 194.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1809-19-4
Molecular Formula C8H19O3P
Molecular Weight (g/mol) 194.21
MDL Number MFCD00066633
InChI Key OSPSWZSRKYCQPF-UHFFFAOYSA-N
Synonym dibutyl phosphite, dibutyl phosphonate, phosphonic acid, dibutyl ester, di-n-butylphosphite, dibutoxyphosphine oxide, mobil dbhp, dibutyl hydrogen phosphonate, di-n-butyl hydrogen phosphite, dibutylfosfit, phosphorous acid, dibutyl ester
PubChem CID 6327349
IUPAC Name dibutoxy(oxo)phosphanium
SMILES CCCCO[P+](=O)OCCCC

Specifications

Density 0.9950g/mL
Boiling Point 118°C to 119°C (11.0 mmHg)
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Refractive Index 1.4225 to 1.4245
Linear Formula [CH3(CH2)3O]2P(O)H
Quantity 100 g
Beilstein 01, I, 187
Specific Gravity 0.995
Solubility Information Solubility in water: slightly soluble
Formula Weight 194.21
Percent Purity 14.5 to 16% (P)
Physical Form Liquid
Chemical Name or Material Dibutyl phosphite, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-316-1
RTECSNumber HS6475000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.