Learn More
Medchemexpress LLC Dibutyl phthalate-d22 | 358731-15-4 | MFCD01861629 | 99.0% | 300.48 | C16D22O4 | 5mg
SDP
Catalog No. 5000268324
Encompass_Preferred
Dibutyl phthalate-d22 is the deuterium labeled Dibutyl phthalate[1] Dibutyl phthalate is a commonly used plasticizer commonly found in some food packaging materials personal care products and the coating of oral medications May cause toxicity and adverse neurobehavioral effects[2][3][4]
Catalog No. 50-002-68324 Supplier Medchemexpress LLC Supplier No. HYY0304S15MG
Dibutyl phthalate-d22 is the deuterium labeled Dibutyl phthalate[1] Dibutyl phthalate is a commonly used plasticizer commonly found in some food packaging materials personal care products and the coating of oral medications May cause toxicity and adverse neurobehavioral effects[2][3][4]