Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibutyl sebacate | 109-43-3 | 98.0% | 314.47 | 5 G
SDP

Catalog No. 5000454562
Encompass_Preferred

Dibutyl sebacate (Dibutyl decanedioate) is a dibutyl ester of sebacic acid, primarily utilized as a plasticizer in the production of plastics.

  • Dibutyl ester of sebacic acid
  • Functions as a plasticizer in plastic production
  • Categorized as a biochemical assay reagent
  • Store at room temperature for 3 years
  • For storage in solvent, store at -80°C for 2 years, or -20°C for 1 year
  • Solubility in DMSO: 100 mg/mL (318.00 mM)
  • In vivo dissolution protocols are available

Catalog No. 50-004-54562 Supplier Medchemexpress LLC Supplier No. HYW0138075G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibutyl sebacate (Dibutyl decanedioate) is a dibutyl ester of sebacic acid, primarily utilized as a plasticizer in the production of plastics.

  • Dibutyl ester of sebacic acid
  • Functions as a plasticizer in plastic production
  • Categorized as a biochemical assay reagent
  • Store at room temperature for 3 years
  • For storage in solvent, store at -80°C for 2 years, or -20°C for 1 year
  • Solubility in DMSO: 100 mg/mL (318.00 mM)
  • In vivo dissolution protocols are available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.