Learn More
Selleck Chemical LLC Dibutyl sebacate
SDP
Catalog No. 5000004215
Encompass_Preferred
Dibutyl sebacate is an organic chemical which is mainly used as a plasticizer in the production of plastics
Catalog No. 50-000-04215 Supplier Selleck Chemical LLC Supplier No. S604125UL
Dibutyl sebacate is an organic chemical which is mainly used as a plasticizer in the production of plastics