Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl succinate, 99%

Catalog No. p-4813455
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453010250 25 mL
AC453011000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453010250 Supplier Thermo Scientific Chemicals Supplier No. 453010250
Only null left
Add to Cart

CAS: 141-03-7 | C12H22O4 | 230.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-03-7
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.3
InChI Key YUXIBTJKHLUKBD-UHFFFAOYSA-N
Synonym dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs
PubChem CID 8830
ChEBI CHEBI:82263
IUPAC Name dibutyl butanedioate
SMILES CCCCOC(=O)CCC(=O)OCCCC

Specifications

Melting Point -29.0°C
Density 0.9800g/mL
Boiling Point 274.0°C
Flash Point 144°C
Assay Percent Range 98.5% min. (GC)
Linear Formula C4H9O2CH2CH2CO2C4H9
Quantity 25 mL
Specific Gravity 0.98
Solubility Information Solubility in water: slightly.
Formula Weight 230.3
Percent Purity 99%
Chemical Name or Material Dibutyl succinate
Show More Show Less
EINECSNumber 205-449-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.