Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl Tin Diacetate, MP Biomedicals

Catalog No. ICN211975 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211975 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211975 Supplier MP Biomedicals, Inc Supplier No. 0521197550

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1067-33-0
Molecular Formula C12H26O4Sn
Molecular Weight (g/mol) 353.05
MDL Number MFCD00008697
InChI Key LDZDQDDROJXUBO-UHFFFAOYSA-N
SMILES CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC

Specifications

Quantity 100 g
Formula Weight 351.02
Chemical Name or Material Dibutyl Tin Diacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.