Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutylamine, 0.04 N in Toluene, Ricca Chemical
SDP

Catalog No. 254921 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
254921 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 254921 Supplier Ricca Chemical Company Supplier No. 2549.21
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-88-3 , 111-92-2
Molecular Formula C8H19N
Molecular Weight (g/mol) Mixture
InChI Key JQVDAXLFBXTEQA-UHFFFAOYSA-N
Synonym dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
PubChem CID 8148
IUPAC Name N-butylbutan-1-amine
SMILES CCCCNCCCC

Specifications

CAS Min % 97.0, 0.58
CAS Max % 100.0, 0.6
Color Colorless to Yellow
Boiling Point Approximately 111°C
Packaging Amber Glass Bottle
Quantity 4 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.04 Normal (N/25) in Toluene
Physical Form Liquid
Chemical Name or Material Dibutylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.