Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyldichlorosilane 94.0+%, TCI America™
SDP

Catalog No. D27045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27045G 5 g
D270425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27045G Supplier TCI America Supplier No. D27045G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3449-28-3
Molecular Formula C8H18Cl2Si
Molecular Weight (g/mol) 213.217
MDL Number MFCD00053202
InChI Key NJKDOKBDBHYMAH-UHFFFAOYSA-N
PubChem CID 76989
IUPAC Name dibutyl(dichloro)silane
SMILES CCCC[Si](CCCC)(Cl)Cl

Specifications

Color Yellow
Boiling Point 109°C
Quantity 5 g
UN Number 2987
Formula Weight 213.22
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyldichlorosilane
EINECSNumber (2)-2041
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.