Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyltin Dichloride 97.0+%, TCI America™
SDP

Catalog No. p-7126093 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D022325G 25 g
D0223100G 100 g
D0223500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D022325G Supplier TCI America Supplier No. D022325G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 683-18-1
Molecular Formula C8H18Cl2Sn
Molecular Weight (g/mol) 303.842
MDL Number MFCD00000518
InChI Key RJGHQTVXGKYATR-UHFFFAOYSA-L
Synonym dibutyltin dichloride, dibutyldichlorotin, di-n-butyltin dichloride, stannane, dibutyldichloro, dibutyltin chloride, dichlorodibutyltin, dibutyl dichloro stannane, dichlorodibutylstannane, dibutyltindichloride, dibutyl tin dichloride
PubChem CID 12688
IUPAC Name dibutyl(dichloro)stannane
SMILES CCCC[Sn](CCCC)(Cl)Cl

Specifications

Melting Point 42°C
Boiling Point 135°C
Quantity 25 g
UN Number 3146
Formula Weight 303.84
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dibutyltin Dichloride
EINECSNumber (2)-2331
RTECSNumber WH7100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.