Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibutyltin Dilaurate

Catalog No. 503081457 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1457 2 kg
29-123-4500G 500 g
21-277-625ML 100 g
29-123-425G 25 g
29-123-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Danger
Catalog No. 50-308-1457 Supplier Sigma Aldrich Supplier No. 2912342KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dibutyltin dilaurate has been used as a catalyst in a protocol for the covalent attachment of poly(ethylene glycol) (PEG) to silicon oxide to form a hydrophilic non-fouling surface. It can also be used as a catalyst in the preparation of polymers by reacting hydroxyl-terminated macromonomers and aliphatic diisocyanates.

Specifications

CAS 77-58-7
Density 1.066 g/mL at 25°C
Molecular Formula C32H64O4Sn
Refractive Index n20/D 1.471
Linear Formula (CH3CH2CH2CH2)2Sn[OCO(CH2)10CH3]2
MDL Number MFCD00008963
Quantity 2 kg
Synonym DBTDL
Molecular Weight (g/mol) 631.56 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.