Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyltin Maleate (so called) [for PVC stabilizer], TCI America™
SDP

Catalog No. D030425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D030425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D030425G Supplier TCI America Supplier No. D030425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78-04-6
Molecular Formula C12H20O4Sn
MDL Number MFCD00014120

Specifications

Quantity 25 g
UN Number 2788
Formula Weight 347.00
Physical Form Liquid
Chemical Name or Material Dibutyltin Maleate (so called) [for PVC stabilizer]
EINECSNumber (2)-2343
RTECSNumber JH4735000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.