Learn More
Enzo Life Sciences Dibutyryl-cAMP, (100mg), CAS Number: 16980-89-5
SDP
Supplier:  Enzo Life Sciences BMLCN1250100
Encompass_Preferred
PKA activator. A cell-permeable cAMP analog which preferentially activates cAMP-dependent protein kinase. The products releaes butyrate due to intracellular and extracellular esterase action. Butyrate was shown to have distinct biological effects. Alternative Name: Dibutyl cyclic AMP sodium salt. Formula: C18H23N5O8P. Na. MW: 491.4. CAS Number: 16980-89-5. Purity: ≥98% (HPLC). Appearance: White crystalline solid. Solubility: Soluble in water (100mg/ml), DMSO (200mM) or 96% ethanol (25mM). Use/Stability: Solution must be stored in the refrigerator and should be lyophilized and frozen for longer storage periods. Long Term Storage: -20°C
Catalog No. NC1539057