Learn More
BIOGEMS INTERNATIONAL INC Dibutyryl-cAMP, sodium salt 250mg
SDP
Catalog No. 501529819
Encompass
Dibutyryl-cAMP is a cell-permeable activator of cAMP-dependent protein kinases and phosphodiesterase inhibitor. It mimics endogenous cyclic adenosine monophosphate (cAMP). Dibutyryl-cAMP is reported to induce apoptosis in vascular smooth muscle cells and induce neurite growth.
Catalog No. 50-152-9819 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 1698950250MG
Dibutyryl-cAMP is a cell-permeable activator of cAMP-dependent protein kinases and phosphodiesterase inhibitor. It mimics endogenous cyclic adenosine monophosphate (cAMP). Dibutyryl-cAMP is reported to induce apoptosis in vascular smooth muscle cells and induce neurite growth.