Learn More
Santa Cruz Biotechnology Dibutyryl-cAMP, 500 mg, Cell-permeable cAMP analog, Phosphodiesterase inhibitor
Supplier:  Santa Cruz Biotechnology SC201567B
Encompass
Dibutyryl-cAMP is a stable analog of cyclic adenosine monophosphate (cAMP), designed to enhance cellular signaling processes. This compound is recognized for its ability to activate cAMP-dependent protein kinases and inhibit phosphodiesterases, which makes it a critical tool in cellular research. Its structure significantly increases lipophilicity, ensuring effective cellular uptake and prolonged action within cells. It is often utilized in studies focused on neural cell differentiation, inflammation modulation, and gene expression alteration.
  • Cell-permeable cAMP analog
  • Potent activator of cAMP-dependent protein kinases
  • Inhibits phosphodiesterases
  • Promotes differentiation and survival of neural stem/progenitor cells
  • Induces neurite outgrowth
  • Exhibits anti-inflammatory activity
  • Promotes wound healing
  • Modulates gene expression
Catalog No. NC1511537