Learn More
STEMCELL Technologies Dibutyryl-cAMP (Sodium Salt), Size: 100 mg
SDP
Catalog No. 501976141
Encompass
Dibutyryl-cAMP is a cell-permeable cyclic AMP (cAMP) analog that activates cAMP-dependent protein kinases (Schwede et al.). This product is supplied as the sodium salt of the molecule.DIFFERENTIATION· Suppresses experimental autoimmune encephalomyelitis development by reducing demyelination and mobilizing neural stem cells in the subventricular zone towards the demyelinated plaques (Khezri et al.).· Induces intrinsic axon growth in peripheral and central nervous systems, and morphological differentiation of astrocytes (Knott et al.; Imamura & Ozawa).· Stimulates neurite outgrowth in PC12 cells (Maruoka et al.).
Catalog No. 50-197-6141 Supplier STEMCELL Technologies Supplier No. 73884
Dibutyryl-cAMP is a cell-permeable cyclic AMP (cAMP) analog that activates cAMP-dependent protein kinases (Schwede et al.). This product is supplied as the sodium salt of the molecule.DIFFERENTIATION· Suppresses experimental autoimmune encephalomyelitis development by reducing demyelination and mobilizing neural stem cells in the subventricular zone towards the demyelinated plaques (Khezri et al.).· Induces intrinsic axon growth in peripheral and central nervous systems, and morphological differentiation of astrocytes (Knott et al.; Imamura & Ozawa).· Stimulates neurite outgrowth in PC12 cells (Maruoka et al.).