Learn More
Enzo Life Sciences Dibutyryl-cGMP (10mg). CAS: 51116-00-8
SDP
Catalog No. 502010453
Encompass_Preferred
A cell-permeable cGMP analog which preferentially activates cGMP-dependent protein kinase. Inhibits thrombin stimulated arachidonic acid release in human platelets. Cell permeable. Purity: ≥96%. Solubility: Soluble in water (100mg/ml). Also soluble in 100% ethanol, DMSO, dimethyl formamide or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0453 Supplier Enzo Life Sciences Supplier No. BMLCN2150010
A cell-permeable cGMP analog which preferentially activates cGMP-dependent protein kinase. Inhibits thrombin stimulated arachidonic acid release in human platelets. Cell permeable. Purity: ≥96%. Solubility: Soluble in water (100mg/ml). Also soluble in 100% ethanol, DMSO, dimethyl formamide or methanol. Long Term Storage: -20°C.