Learn More
Medchemexpress LLC Dibutyryl-cGMP sodium | 51116-00-8 | 99.7% | 100 MG
SDP
Catalog No. 5000340919
Encompass_Preferred
Dibutyryl-cGMP sodium is a cell-permeable cGMP analogue. It preferentially activates cGMP-dependent protein kinase (PKG), inhibits the release of [3H]-arachidonic acid from γ thrombin-stimulated human platelets, and induces peripheral antinociception through ATP-sensitive K+ channel activation.
  • Cell-permeable cGMP analogue
  • Preferentially activates cGMP-dependent protein kinase (PKG)
  • Inhibits release of [3H]-arachidonic acid from γ thrombin-stimulated human platelets
  • Induces peripheral antinociception via activation of ATP-sensitive K+ channels
  • Induces process elongation and branching in astrocytes
  • Prevents stress fibre formation and induces stellate morphology in cerebellar astrocytes
  • Prevents RhoA-membrane association
  • Accelerates wound closure in scratchwound model
  • Antagonizes the hyperalgesic effect of PGE2
Catalog No. 50-003-40919 Supplier Medchemexpress LLC Supplier No. HY130354100MG
Dibutyryl-cGMP sodium is a cell-permeable cGMP analogue. It preferentially activates cGMP-dependent protein kinase (PKG), inhibits the release of [3H]-arachidonic acid from γ thrombin-stimulated human platelets, and induces peripheral antinociception through ATP-sensitive K+ channel activation.
  • Cell-permeable cGMP analogue
  • Preferentially activates cGMP-dependent protein kinase (PKG)
  • Inhibits release of [3H]-arachidonic acid from γ thrombin-stimulated human platelets
  • Induces peripheral antinociception via activation of ATP-sensitive K+ channels
  • Induces process elongation and branching in astrocytes
  • Prevents stress fibre formation and induces stellate morphology in cerebellar astrocytes
  • Prevents RhoA-membrane association
  • Accelerates wound closure in scratchwound model
  • Antagonizes the hyperalgesic effect of PGE2