Learn More
Cayman Chemical Dibutyryl-Cyclic GMP (sodium salt), Purity: ≥98%, CAS: 51116-00-8, 50 mg
SDP
Catalog No. 502968893
Encompass_Preferred
A cell-permeable, cGMP analog that activates cGMP-dependent protein kinase
Catalog No. 50-296-8893 Supplier Cayman Chemical Supplier No. 1599150 MG
A cell-permeable, cGMP analog that activates cGMP-dependent protein kinase