Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dic

Catalog No. D1254075G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1254075G 5 g
D125407100G 100 g
D125407500G 500 g
D12540725G 25 g
D1254071KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Danger
Catalog No. D1254075G Supplier Sigma Aldrich Supplier No. D1254075G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DIC is an abbreviated form of N,N'sa-diisopropylcarbodiimide. It plays a role in the cyclization of N-(+a-hydroxy)amides to form 2-oxazolines.DIC (N,N'sa-Diisopropylcarbodiimide) has been used in combination with 1-hydroxy-7-azabenzotriazole (HOAt) for the coupling of amino acid with N-allylglycine to form N-allylpeptide.

Specifications

CAS 693-13-0
Density 0.815 g/mL at 20°C
Boiling Point 145°C to 148°C
Molecular Formula C7H14N2
Refractive Index n20/D 1.433
Linear Formula (CH3)2CHN=C=NCH(CH3)2
MDL Number MFCD00065689
Quantity 5 g
Synonym N,N'-Diisopropylcarbodiimide
Molecular Weight (g/mol) 126.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.