Learn More
Sigma Aldrich Fine Chemicals Biosciences DIC 991KG diisopropylcarbodiim
Catalog No. 502906989
Encompass
DIC is an abbreviated form of N NAE-diisopropylcarbodiimide. It plays a role in the cyclization of N-(-hydroxy)amides to form 2-oxazolines.
Catalog No. 50-290-6989 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D1254071KG
DIC is an abbreviated form of N NAE-diisopropylcarbodiimide. It plays a role in the cyclization of N-(-hydroxy)amides to form 2-oxazolines.