Learn More
Ibidi USA DIC Lid for µ-Dishes
SDP
Catalog No. NC3073854
Encompass
Special lid with a glass coverslip insert for use in Differential Interference Contrast (DIC) microscopy. Ideal for use in Differential Interference Contrast (DIC) microscopy. Suitable for use with all ibidi 35 mm µ-Dishes. Can be alcohol-sterilized and then be reused
Catalog No. NC3073854 Supplier Ibidi USA Supplier No. 80051
Special lid with a glass coverslip insert for use in Differential Interference Contrast (DIC) microscopy. Ideal for use in Differential Interference Contrast (DIC) microscopy. Suitable for use with all ibidi 35 mm µ-Dishes. Can be alcohol-sterilized and then be reused