Learn More
Ibidi USA Specialty Lid for Differential interference contrast (DIC) microscopy, 15 count
SDP
Catalog No. NC2079774
Encompass
Special lid for use in Differential interference contrast (DIC) microscopy. Suitable for use with all ibidi µ-Slide (except channel and PH+ versions). Made from special low birefringence polymer. Pcs./Box: 15 (individually packed)
This item is not returnable. View return policy
Catalog No. NC2079774 Supplier Ibidi USA Supplier No. 80055
Special lid for use in Differential interference contrast (DIC) microscopy. Suitable for use with all ibidi µ-Slide (except channel and PH+ versions). Made from special low birefringence polymer. Pcs./Box: 15 (individually packed)