Learn More
Bioworld DICAMBA, 250 mg
Supplier:  Bioworld 404500161
Encompass_Preferred
DicambaSynonyms: 3,6-Dichloro-o-anisic acidAuxin like growth regulatorA herbicide derived from benzoic acid Melting Point: 112-116 °C(lit.)
Catalog No. 50-255-888