Learn More
Crescent Chemical Co Inc DICAMBA-5-HYDROXY SOLN 1 ML
SDP
Catalog No. 5080187
Encompass_Preferred
Dicamba-5-hydroxy 100 µg/mL in Acetonitrile Herbicide (Metabolite solution) CAS 7600-50-2 Molecular Weight - 237.0368 Storage - RT Catalog Num - DRE-XA12260500AL pesticide
Catalog No. 50-801-87 Supplier Crescent Chemical Co Inc Supplier No. PSXA12260500AL
Dicamba-5-hydroxy 100 µg/mL in Acetonitrile Herbicide (Metabolite solution) CAS 7600-50-2 Molecular Weight - 237.0368 Storage - RT Catalog Num - DRE-XA12260500AL pesticide