Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicarbonyl(2,4-pentanedionato)rhodium(I), 97%

Catalog No. AA39295MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39295MD 250 mg
AA3929503 1 g
AA3929506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA39295MD Supplier Thermo Scientific Chemicals Supplier No. 039295MD
Add to Cart

CAS: 14874-82-9 | C7H7O4Rh | 258.03 g/mol

(Acetylacetonato)dicarbonylrhodium(I) is used in in situ formation of a floors-soluble hydroformylation catalyst of interest in molecular engineering. It is also used as a catalyst for various carbonylation reactions, silylcarbocyclizations, conjugate additions to eons, carbamoylstannation, and reduction of aromatic nitro compounds. It is also used in Hydroformylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14874-82-9
Molecular Formula C7H7O4Rh
Molecular Weight (g/mol) 258.03
MDL Number MFCD00009884
InChI Key BZCAWKOPWNIDOC-FGSKAQBVSA-M
Synonym (Acetylacetonato)dicarbonylrhodium(I), Acetylacetonatorhodium(I) dicarbonyl
IUPAC Name λ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate
SMILES [Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O

Specifications

Melting Point 144°C to 147° (sublimation)
Color Green
Odor Odorless
Assay Percent Range 99%
Quantity 250 mg
Solubility Information Soluble in acetone
Formula Weight 258.04
Physical Form Crystalline
Chemical Name or Material Dicarbonyl(2, 4-pentanedionato)rhodium(I)
Hazard Category H228-H301-H317-H319
Precautionary Statement P210-P240-P241-P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 6.1; Packaging Group: III
EINECSNumber 238-947-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.