Learn More
Strem, An Ascensus Company CAS# 32627-01-3. 100mg. Dicarbonyl(pentamethylcyclopentadienyl)rhodium(I), 99% (99.9%-Rh). MFCD00058847
SDP
Supplier:  Strem, An Ascensus Company 450739100MG
Encompass
CAS# 32627-01-3. 100mg. Dicarbonyl(pentamethylcyclopentadienyl)rhodium(I), 99% (99.9%-Rh). MFCD00058847. Molecular Weight: 294.16. Molecular Formula: (CH3)5C5Rh(CO)2. Color/form: red xtl. Strem# 45-0739. http://www.strem.com/catalog/v/45-0739/
Catalog No. 50-901-12178