Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dicarbonylcyclopentadienyl Cobalt(I)

Catalog No. 24525925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-525-925G 25 g
24-525-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 24-525-925G Supplier Sigma Aldrich Supplier No. 24525925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mediates the [2+2+2]-cyclization of allenediynes to tricyclic systems.

Specifications

CAS 12078-25-0
Boiling Point 139°C to 140°C (710 mmHg, lit.)
Molecular Formula C7H5CoO2
Linear Formula C5H5Co(CO)2
MDL Number MFCD00009589
Quantity 25 g
Synonym CpCo(CO)2; Cobalt dicarbonyl cyclopentadiene; Cyclopentadienyl cobalt(I) dicarbonyl; Cyclopentadienylcobalt dicarbonyl; Cyclopentadienyldicarbonylcobalt
Molecular Weight (g/mol) 180.05 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.