Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicarbonylcyclopentadienylcobalt(I) 95.0+%, TCI America™
SDP

Catalog No. D49402G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49402G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D49402G Supplier TCI America Supplier No. D49402G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12078-25-0
Molecular Formula C7H5CoO2
MDL Number MFCD00009589
Synonym Cyclopentadienylcobalt Dicarbonyl

Specifications

Melting Point -22°C
Boiling Point 140°C
Quantity 2 g
UN Number 1992
Formula Weight 180.05
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Dicarbonylcyclopentadienylcobalt(I)
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.