Learn More
Description
Use these double dice to teach probability, predict numerical outcomes and track the statistical results of repeated rolls.
- Each brightly colored, translucent outer die holds a single, smaller, white die inside
- Twice the dice means twice the math and twice the learning
- Set of 72 dice comes in a sturdy hexagonal tub for quick clean-up and easy storage
Order Info
Grades 1+
Specifications
Specifications
| Color | Assorted |
| Grade Level | Elementary |
| Grade | 1 |
| For Use With (Application) | Teach probability, predict numerical outcomes and track the statistical results of repeated rolls |
| Includes | 72 dice comes in a sturdy hexagonal tub |
| Description | Double dice |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
