Learn More
Medchemexpress LLC Dicentrine | 517-66-8 | 5 MG
SDP
Catalog No. 5000418426
Encompass_Preferred
Dicentrine is a natural product isolated from the plant Stephania epigaea Lo that exhibits an antihypertensive effect. It functions as an α1-adrenoceptor antagonist and has been shown to be effective against human hyperplastic prostates.
  • Isolated from the plant Stephania epigaea Lo.
  • Exhibits an antihypertensive effect.
  • Functions as an α1-adrenoceptor antagonist.
  • Effective against human hyperplastic prostates.
  • For research use only.
  • High purity (99.94%).
Catalog No. 50-004-18426 Supplier Medchemexpress LLC Supplier No. HYN69695MG
Dicentrine is a natural product isolated from the plant Stephania epigaea Lo that exhibits an antihypertensive effect. It functions as an α1-adrenoceptor antagonist and has been shown to be effective against human hyperplastic prostates.
  • Isolated from the plant Stephania epigaea Lo.
  • Exhibits an antihypertensive effect.
  • Functions as an α1-adrenoceptor antagonist.
  • Effective against human hyperplastic prostates.
  • For research use only.
  • High purity (99.94%).