Learn More
BPS Bioscience Inc Human Dicer, His-Tag, 10ug
SDP
Supplier:  BPS Bioscience Inc 790831
Encompass
Human Dicer, also known as Dicer 1, Ribonuclease III, GenBank Accession No. NM_030621, a.a. 2-1992 (end), with a N-terminal His-Tag, expressed in an Sf9 cell expression system. MW =221 kDa. 10ug; BPS BIOSCIENCE INC, mfg# 79083-1
Catalog No. NC1853383