Learn More
TARGETMOL CHEMICALS INC Dichlorisone Acetate 50MG
SDP
Catalog No. 502880718
Encompass_Preferred
Also available in 1 mL, 10 mg and bulk. Please contact Fisher for quotes. Dichlorisone Acetate (Diloderm) is a synthetic glucocorticoid corticosteroid with anti-inflammatory activity. Purity 99.72%
Catalog No. 50-288-0718 Supplier TARGETMOL CHEMICALS INC Supplier No. T026950MG
Also available in 1 mL, 10 mg and bulk. Please contact Fisher for quotes. Dichlorisone Acetate (Diloderm) is a synthetic glucocorticoid corticosteroid with anti-inflammatory activity. Purity 99.72%