Learn More
Strem, An Ascensus Company CAS# 95408-45-0. Dichloro[1,1'-bis(di-t-butylphosphino)ferrocene]palladium(II), 99%. MFCD08064219. 500MG
SDP
Supplier:  Strem, An Ascensus Company 460445500MG
Encompass
CAS# 95408-45-0. 500MG. Dichloro[1,1'-bis(di-t-butylphosphino)ferrocene]palladium(II), 99%. MFCD08064219. Molecular Formula: C26H44Cl2FeP2Pd. Molecular Weight: 651.75. Color/Form: brown to purple xtl. Strem#: 46-0445 www.strem.com/catalog/v3/46-0445/
Catalog No. 50-901-12341