Learn More
Strem, An Ascensus Company CAS# 95464-05-4. Dichloro 1,1'-bis(diphenylphosphino)ferrocene palladium (II) dichloromethane, 99%. MFCD00792899. 1G.
SDP
Supplier:  Strem, An Ascensus Company 4604501G
Encompass
CAS# 95464-05-4. 1G. Dichloro 1,1'-bis(diphenylphosphino)ferrocene palladium (II) dichloromethane, 99%. MFCD00792899. Molecular Formula: C35H30Cl4FeP2Pd. Molecular Weight: 731.77 (816.65). Color/form orange-red pwdr. Strem#: 46-0450. www.strem.com/catalog/v3/46-0450/
Catalog No. 50-901-12343