Learn More
Medchemexpress LLC Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane adduct | 95464-05-4 | 98.5% | 816.64 | 10 G
SDP
Supplier:  Medchemexpress LLC HY2162310G
Encompass_Preferred
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane adduct is a common dichloromethane solvate of the palladium catalyst Pd(dppf)Cl2, a phosphine ligand palladium(II) complex. It serves as a versatile precatalyst for various cross-coupling reactions, stabilizing reaction intermediates and lowering activation energy. This adduct is suitable for modifying complex bioactive molecules containing free amino groups, eliminating the need to change catalysts for different reaction types. It is intended for research use only and is a solid with an orange to red color.
  • Versatile precatalyst for cross-coupling reactions
  • Stabilizes reaction intermediates
  • Lowers reaction activation energy
  • Can be used in Suzuki-Miyaura coupling reaction studies
  • Suitable for modifying complex bioactive molecules containing free amino groups
  • Purifiable by column chromatography after reaction
Catalog No. 50-004-00845