Learn More
Medchemexpress LLC Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane adduct | 95464-05-4 | 98.5% | 816.64 | 100 G
SDP
Catalog No. 5000402621
Encompass_Preferred
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane adduct is a common dichloromethane solvate of the palladium catalyst Pd(dppf)Cl2, a phosphine ligand palladium(II) complex. It serves as a versatile precatalyst for various cross-coupling reactions.
  • Stabilizes reaction intermediates and lowers reaction activation energy in coupling reactions.
  • Useful in Suzuki-Miyaura coupling reaction studies.
  • Efficiently catalyzes various cross-coupling reactions.
  • Suitable for modifying complex bioactive molecules containing free amino groups.
  • Product can be purified by column chromatography before biological activity testing.
Catalog No. 50-004-02621 Supplier Medchemexpress LLC Supplier No. HY21623100G
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane adduct is a common dichloromethane solvate of the palladium catalyst Pd(dppf)Cl2, a phosphine ligand palladium(II) complex. It serves as a versatile precatalyst for various cross-coupling reactions.
  • Stabilizes reaction intermediates and lowers reaction activation energy in coupling reactions.
  • Useful in Suzuki-Miyaura coupling reaction studies.
  • Efficiently catalyzes various cross-coupling reactions.
  • Suitable for modifying complex bioactive molecules containing free amino groups.
  • Product can be purified by column chromatography before biological activity testing.