Learn More
Medchemexpress LLC Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane adduct | 95464-05-4 | 98.5% | 816.64 | 50 G
SDP
Catalog No. 5000400763
Encompass_Preferred
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane adduct is a common dichloromethane solvate of the palladium catalyst Pd(dppf)Cl2. This phosphine ligand palladium(II) complex serves as a versatile precatalyst for various cross-coupling reactions. It efficiently catalyzes various cross-coupling reactions and is suitable for modifying complex bioactive molecules containing free amino groups, eliminating the need to change catalysts for different reaction types.
  • Stabilizes reaction intermediates
  • Lowers reaction activation energy in coupling reactions
  • Used in Suzuki-Miyaura coupling reaction studies
  • Product can be purified by column chromatography before biological activity testing
Catalog No. 50-004-00763 Supplier Medchemexpress LLC Supplier No. HY2162350G
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane adduct is a common dichloromethane solvate of the palladium catalyst Pd(dppf)Cl2. This phosphine ligand palladium(II) complex serves as a versatile precatalyst for various cross-coupling reactions. It efficiently catalyzes various cross-coupling reactions and is suitable for modifying complex bioactive molecules containing free amino groups, eliminating the need to change catalysts for different reaction types.
  • Stabilizes reaction intermediates
  • Lowers reaction activation energy in coupling reactions
  • Used in Suzuki-Miyaura coupling reaction studies
  • Product can be purified by column chromatography before biological activity testing