Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dichloro(1,10-Phenanthroline)Palladium(II)

Catalog No. 3987991G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-879-91G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-879-91G Supplier Sigma Aldrich Supplier No. 3987991G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for:Carbonylation of alkyl nitritesArylation reactionsMultiple acetylene insertions under Sonogashira reaction protocolCross-coupling reactions of pyridine carboxylic acid chlorides with alkylzinc reagentsHydrophosphinylation of alkenes and alkynesHeck reactionRegioselective and stereoselective aminohalogenation reactionsReaction of chloroenynes and chlorodienes with Grignard reagents for the synthesis of stereodefined enynes and dienesAffects the aggregation of human prion protein PrP106-126

Specifications

CAS 500996-04-3
Molecular Formula C11H13BF4N2
Linear Formula C11H13BF4N2
MDL Number MFCD07784449
Quantity 50 g
Molecular Weight (g/mol) 260.04 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.