Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dichloro(1,2-diaminocyclohexane)platinum(II) | 52691-24-4 | MFCD00058826 | 98.0% | 380.17 g·mol⁻¹ | C6H14Cl2N2Pt | 500 MG
SDP

Catalog No. 5000281220
Encompass_Preferred

Dichloro(1,2-diaminocyclohexane)platinum(II) is a platinum(II) dichloro complex provided as a research-grade biochemical reagent for chemical and biological studies of platinum-based compounds. It is commonly used as a reference compound or intermediate in synthesis and in studies of platinum antiproliferative activity.

  • Molecular formula: C6H14Cl2N2Pt.
  • Molecular weight: 380.17 g·mol⁻¹.
  • CAS number: 52691-24-4.
  • Typical purity around 98.0%.
  • Available as a 500 mg quantity.
  • Suitable for chemical research and analytical applications.

Catalog No. 50-002-81220 Supplier Medchemexpress LLC Supplier No. HYW414604500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dichloro(1,2-diaminocyclohexane)platinum(II) is a platinum(II) dichloro complex provided as a research-grade biochemical reagent for chemical and biological studies of platinum-based compounds. It is commonly used as a reference compound or intermediate in synthesis and in studies of platinum antiproliferative activity.

  • Molecular formula: C6H14Cl2N2Pt.
  • Molecular weight: 380.17 g·mol⁻¹.
  • CAS number: 52691-24-4.
  • Typical purity around 98.0%.
  • Available as a 500 mg quantity.
  • Suitable for chemical research and analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.