Learn More
Medchemexpress LLC Dichloro(1,2-diaminocyclohexane)platinum(II) | 52691-24-4 | MFCD00058826 | 98.0% | 380.18 g/mol | C6H14Cl2N2Pt | 250 MG

Supplier: Medchemexpress LLC HYW414604250MG
Dichloro(1,2-diaminocyclohexane)platinum(II) is a platinum(II) complex used as a research reagent. Key identifiers include CAS 52691-24-4, molecular formula C6H14Cl2N2Pt, and molecular weight 380.18 g/mol. Manufacturer documentation provides storage guidance for both solid material and solutions.
- Platinum(II) complex for research applications.
- Molecular formula C6H14Cl2N2Pt.
- Molecular weight 380.18 g/mol.
- CAS number 52691-24-4.
- Storage: RT under nitrogen; in solvent, store at -80°C up to 2 years or -20°C up to 1 year.
- Supplied in milligram quantities suitable for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.