Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dichloro[1,3-Bis(2,4,6-Trimethylphenyl)-2-Imidazolidinylidene][[5-[(Dimethylamino)Sulfonyl]-2-(1-Methylethoxy-O)Phenyl]Methylene-C]Ruthenium(II)

Catalog No. 762261500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-226-1500MG 500 mg
76-226-12G 2 g
76-226-1100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 76-226-1500MG Supplier Sigma Aldrich Supplier No. 762261500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zhan Catalyst-1B (Hoveyda-Grubbs type catalyst) can be used:In the preparation of hybrid olefin metathesis catalysts, with increased catalytic activity via immobilization on molecular seives. supportsAs a catalyst in the synthesis of divergent dendrimerIn ring-closure metathesis (RCM) reaction.

Specifications

CAS 918870-76-5
Molecular Formula C33H43Cl2N3O3RuS
Linear Formula C33H43Cl2N3O3RuS
MDL Number MFCD08459353
Quantity 500 mg
Synonym Zhan Catalyst-1B
Molecular Weight (g/mol) 733.75 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.