Learn More
Strem, An Ascensus Company CAS# 59831-02-6. 1g. Dichloro(1,3-bis(diphenylphosphino)propane)palladium(II), 98%. MFCD03844773
SDP
Catalog No. 5090112357
Encompass
CAS# 59831-02-6. 1g. Dichloro(1,3-bis(diphenylphosphino)propane)palladium(II), 98%. MFCD03844773. Molecular Weight: 589.77. Molecular Formula: PdCl2[(C6H5)2PCH2CH2CH2P(C6H5)2]. Color/form: pale yellow pwdr. Strem# 46-0850. http://www.strem.com/catalog/v/46-0850/
Catalog No. 50-901-12357 Supplier Strem, An Ascensus Company Supplier No. 4608501G
CAS# 59831-02-6. 1g. Dichloro(1,3-bis(diphenylphosphino)propane)palladium(II), 98%. MFCD03844773. Molecular Weight: 589.77. Molecular Formula: PdCl2[(C6H5)2PCH2CH2CH2P(C6H5)2]. Color/form: pale yellow pwdr. Strem# 46-0850. http://www.strem.com/catalog/v/46-0850/