Learn More
Strem, An Ascensus Company CAS# 205319-10-4. Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), min. 98% MFCD14155707. 1G
SDP
Catalog No. 501385535
Encompass
CAS# 205319-10-4. 1G. Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), min. 98%. MFCD14155707. Molecular Formula: C39H32Cl2OP2Pd. Molecular Weight: 755.94. Color/Form: yellow microxtls. Strem#: 46-0865 www.strem.com/catalog/v3/46-0865/
Catalog No. 50-138-5535 Supplier Strem, An Ascensus Company Supplier No. 4608651G
CAS# 205319-10-4. 1G. Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), min. 98%. MFCD14155707. Molecular Formula: C39H32Cl2OP2Pd. Molecular Weight: 755.94. Color/Form: yellow microxtls. Strem#: 46-0865 www.strem.com/catalog/v3/46-0865/