Learn More
Medchemexpress LLC Dichloro(benzene)ruthenium(II) dimer | 37366-09-9 | MFCD00064686 | 98.0% | 500.18 | C12H12Cl4Ru2 | 500 MG
SDP
Catalog No. 5000299406
Encompass_Preferred
Dichloro(benzene)ruthenium(II) dimer is an organometallic reagent and catalyst precursor used in synthetic and homogeneous catalytic applications. It is supplied as a brown to reddish-brown solid with high reported purity and defined physicochemical properties, and is intended for research use under controlled laboratory conditions.
  • Organometallic reagent for synthesis and catalysis.
  • Brown to reddish-brown solid with high purity (98.0%).
  • Molecular formula C12H12Cl4Ru2 and molecular weight 500.18.
  • Stable when stored protected from light and under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
  • Available in multiple pack sizes from 100 mg to 10 g.
  • Suitable as a catalyst precursor or reagent in organometallic research.
Catalog No. 50-002-99406 Supplier Medchemexpress LLC Supplier No. HYW033466500MG
Dichloro(benzene)ruthenium(II) dimer is an organometallic reagent and catalyst precursor used in synthetic and homogeneous catalytic applications. It is supplied as a brown to reddish-brown solid with high reported purity and defined physicochemical properties, and is intended for research use under controlled laboratory conditions.
  • Organometallic reagent for synthesis and catalysis.
  • Brown to reddish-brown solid with high purity (98.0%).
  • Molecular formula C12H12Cl4Ru2 and molecular weight 500.18.
  • Stable when stored protected from light and under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
  • Available in multiple pack sizes from 100 mg to 10 g.
  • Suitable as a catalyst precursor or reagent in organometallic research.