Learn More
Medchemexpress LLC Dichloro(benzene)ruthenium(II) dimer | 37366-09-9 | MFCD00064686 | 98.0% | 500.18 g·mol⁻¹ | C12H12Cl4Ru2 | 100 MG
SDP
Catalog No. 5000299566
Encompass_Preferred
Dichloro(benzene)ruthenium(II) dimer is an organometallic reagent used as a catalyst precursor and synthetic building block in organometallic and synthetic chemistry research. This red-orange, diamagnetic solid is supplied with high purity for laboratory use and is commonly used in catalyst development, mechanistic studies, and the synthesis of ruthenium complexes.
  • High purity (98.0%) for reproducible experimental results.
  • Well-characterized molecular formula C12H12Cl4Ru2 and molecular weight 500.18 g·mol⁻¹.
  • Suitable as a catalyst precursor in organometallic synthesis.
  • Available in multiple pack sizes to support different experimental scales.
  • Useful for mechanistic studies and preparation of coordination complexes.
Catalog No. 50-002-99566 Supplier Medchemexpress LLC Supplier No. HYW033466100MG
Dichloro(benzene)ruthenium(II) dimer is an organometallic reagent used as a catalyst precursor and synthetic building block in organometallic and synthetic chemistry research. This red-orange, diamagnetic solid is supplied with high purity for laboratory use and is commonly used in catalyst development, mechanistic studies, and the synthesis of ruthenium complexes.
  • High purity (98.0%) for reproducible experimental results.
  • Well-characterized molecular formula C12H12Cl4Ru2 and molecular weight 500.18 g·mol⁻¹.
  • Suitable as a catalyst precursor in organometallic synthesis.
  • Available in multiple pack sizes to support different experimental scales.
  • Useful for mechanistic studies and preparation of coordination complexes.